missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, tribasic, anhydrous, tech., Thermo Scientific Chemicals

Catalog Number p-4407929
Click to view available options
Quantity:
1 kg
5 kg
7601-54-9
Na3O4P
163.939
MFCD00003510
RYFMWSXOAZQYPI-UHFFFAOYSA-K
trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
24243
CHEBI:37583
trisodium;phosphate
[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
7601-54-9
Na3O4P
163.939
MFCD00003510
RYFMWSXOAZQYPI-UHFFFAOYSA-K
trisodium phosphate, sodium orthophosphate, tromete, tribasic sodium phosphate, phosphoric acid, trisodium salt, oakite, sodium phosphate, anhydrous, trisodium orthophosphate, phosphoric acid trisodium salt, trinatriumphosphat
24243
CHEBI:37583
trisodium;phosphate
[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Sodium phosphate, tribasic is used in soaps, detergents, photographic developers, water softeners and boiler scale removers. It is used as a food additive, as cleaning agents, as a protein precipitant and an active ingredient in fluxes designed to deoxygenate nonferrous metals for casting. It is used to prepare chlorinated trisodium phosphate by reacting with sodium hypochlorite and used as a disinfectant and bleach. Further, it is used in dairy substitutes and to inhibit mold. In addition, it is employed in ceramic production. It is also used for the preservation of biologically active soil samples and fractionization of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 1 kg
Solubility Information Soluble in water. Insoluble in ethanol and carbon disulfide.
Formula Weight 163.94
Grade Technical
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder
Density 1.62g/cm3
Chemical Name or Material Sodium phosphate, tribasic, anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Sodium phosphate, tribasic, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : trisodium phosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'