missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
121.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
For tissue culture and molecular biology applications
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.
Specifications
Boiling Point | 203.9°C |
Melting Point | 100°C |
Color | White |
pH | 4.0 - 6.0 |
Physical Form | Solid |
Quantity | 1 kg |
Assay Percent Range | ≥99 % |
Merck Index | 15, 8790 |
IUPAC Name | sodium dihydrogen phosphate |
Formula Weight | 119.98 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'