missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic, 98%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4813657
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
7558-80-7
H2NaO4P
119.98
MFCD00003527,MFCD00146206
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
23672064
CHEBI:37585
sodium;dihydrogen phosphate
[Na+].OP(O)([O-])=O
7558-80-7
H2NaO4P
119.98
MFCD00003527,MFCD00146206
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
23672064
CHEBI:37585
sodium;dihydrogen phosphate
[Na+].OP(O)([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 kg
Assay Percent Range 98%
Linear Formula NaH2PO4
Merck Index 15,879
Solubility Information Solubility in water: 850g/L (20°C).
Formula Weight 119.98
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'