missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic dodecahydrate, meets analytical spec. of BP, Ph. Eur., E339, 98.5-101% (calc. to the dried substance), Honeywell Fluka™

Brand:  Honeywell Fluka 04273-25KG

Code : NEW

Additional Details : CAS Number : 10039-32-4 Weight : 25.00000kg

Quantity 25kg

 View more versions of this product

Product Code. 15640750

  • £150.43 / 25kg


Estimated Shipment date
from Supplier 26-01-2018
Add to basket

Specifications

Chemical Identifiers

10039-32-4
98.5 to 101% (calc. to the dried substance)
Na2HPO4 · 12H2O
meets analytical specification of Ph.Eur., BP, E339, 98.5-101% calc. to the dried substance
Sodium phosphate dibasic dodecahydrate
358.14
MFCD00149181

Specifications

Sodium phosphate dibasic dodecahydrate
10039-32-4
Reag. Ph. Eur.
98.5 to 101%
Na2HPO4 · 12H2O
MFCD00149181
max. 1 ppm
Can
meets analytical specification of Ph.Eur., BP, E339, 98.5-101% calc. to the dried substance
100.0000%
98.5 to 101% (calc. to the dried substance)
assay (calc. to the dried substance)
358.14
max. 10 ppm
max. 61%
25kg
Videos