missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, Anhydrous, U.S.P. - F.C.C., J.T. Baker™
180.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Quantity | 500 g |
Formula Weight | 141.96 |
Grade | FCC/USP |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 2.07kg |
Chemical Name or Material | Sodium Phosphate, Dibasic, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'