missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5383217
246.05 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
[Na+].[Na+].OP([O-])([O-])=O

High quality chemicals for laboratory and specialized industrial use

    Suitable for Buffer Solutions
Quantity 500 g
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96
Grade Meets ACS reagent chemical requirements
Packaging Poly Bottle
Specific Gravity 1L = 2.07kg
Chemical Name or Material Sodium Phosphate Dibasic, Anhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'