missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, 7-Hydrate, Crystal, U.S.P., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Specifications
Physical Form | Crystals |
Quantity | 500 g |
Molecular Formula | H17Na2O11P |
Molecular Weight (g/mol) | 270.08 |
Formula Weight | 268.07 |
Grade | USP |
Packaging | Wide Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.70kg |
Chemical Name or Material | Sodium Phosphate, Dibasic, 7-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'