missing translation for 'onlineSavingsMsg'
Learn More

Sodium persulfate, +98%, Thermo Scientific Chemicals

Catalog Number 10506631
95.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
7775-27-1
Na2O8S2
238.092
MFCD00003501
CHQMHPLRPQMAMX-UHFFFAOYSA-L
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
62655
disodium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
7775-27-1
Na2O8S2
238.092
MFCD00003501
CHQMHPLRPQMAMX-UHFFFAOYSA-L
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
62655
disodium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Color White to Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Assay Percent Range 98+%
Linear Formula Na2S2O8
Fieser 01,1102; 03,267
Merck Index 15, 8787
Formula Weight 238.09
Percent Purity ≥98.0% (Iodometry)
Identification Pass Test
Packaging Plastic bottle
Chemical Name or Material Sodium persulfate, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Persulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'