missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perrhenate, 99+% (metals basis), Re 68%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 300°C |
Color | Yellow |
Physical Form | Crystalline |
Quantity | 10 g |
Assay Percent Range | 99+% (metals basis) |
Solubility Information | Soluble in water and ethanol. |
Formula Weight | 273.19 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystalline powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'