missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxide, ACS reagent, ACROS Organics™

Brand:  Acros Organics 315761000

Code : 29

Additional Details : CAS Number : 1313-60-6 Weight : 0.15000kg Transport : UN number : 1504 Chem class : 1 Pack group : I


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10678411

  • £84.70 / 100g


EU Stock 11
Add to basket


Specifications

Chemical Identifiers

Sodium peroxide
ACS Reagent Grade
Na2O2
77.97
1313-60-6
≥93.0%
Na2O2
MFCD00003497

Specifications

Sodium peroxide
ACS Reagent Grade
≥93.0%
Na2O2
MFCD00003497
15, 8786
2.8000g/mL
50ppm max.
5ppm max.
2.8
Yellow
100g
1313-60-6
Powder or Granules
Na2O2
77.97
06,548; 07,341; 11,492
20ppm max.
20ppm max.
Plastic bottle
Solubility in water: reacts violently. Other solubilities: forming naoh and h2o2, freely soluble
10ppm max.
460.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium peroxide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • H271-May cause fire or explosion; strong oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS