missing translation for 'onlineSavingsMsg'
Learn More

Sodium periodate, 99%, for analysis, ACROS Organics™

Brand:  Acros Organics 198380025

Code : 29

Additional Details : CAS Number : 7790-28-5 Weight : 2.55000kg Transport : UN number : 3085 Chem class : 1a Pack group : I


Quantity 2.5kg
Packaging Plastic Bottle

 View more versions of this product

Product Code. 11934221

  • £668.40 / 2.50kg


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Sodium Periodate
For analysis
99%
NaIO4
7790-28-5
99%
INaO4
213.89

Specifications

Sodium Periodate
For analysis
99%
INaO4
213.89
300ppm max.
10ppm max.
2000ppm max.
Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents
White to almost white
3.4 to 4.5 (5% soln. at 20°C)
7790-28-5
Fine Crystals
99%
NaIO4
01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294
10ppm max.
20ppm max.
Plastic Bottle
200ppm max.
300.0°C
2.5kg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium periodate

Signal Word
  • Danger

Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H271-May cause fire or explosion; strong oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P221-Take any precaution to avoid mixing with combustibles A§.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS