missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3735062
66.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7601-89-0
ClNaO4
122.44
MFCD00003480
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
522606
sodium;perchlorate
[O-]Cl(=O)(=O)=O.[Na+]
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7601-89-0
ClNaO4
122.44
MFCD00003480
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
522606
sodium;perchlorate
[O-]Cl(=O)(=O)=O.[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 482.0°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 250 g
Assay Percent Range 98%
Linear Formula NaClO4
Merck Index 15, 8784
Solubility Information Solubility in water: 209 g/100mL (15°C). Other solubilities: 170 g/100mL water (0°C), 300 g/100mL water (80°C), soluble in alcohol
Formula Weight 122.44
Percent Purity 98%
Grade Extra Pure
Loss on Drying 2 % max. (130°C, 2 hrs)
Packaging Plastic bottle
Chemical Name or Material Sodium perchlorate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'