missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perchlorate, 98%, extra pure, Thermo Scientific Chemicals
344.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 482.0°C |
Color | Colorless to Yellow |
Physical Form | Liquid |
Quantity | 2.5 kg |
Assay Percent Range | 98% |
Linear Formula | NaClO4 |
Merck Index | 15, 8784 |
Solubility Information | Solubility in water: 209g/100mL (15°C). Other solubilities: 170g/100mL water (0°C),300g/100mL water (80°C),soluble in alcohol |
Formula Weight | 122.44 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'