missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Color | White |
Physical Form | Granular Powder |
Quantity | 10 kg |
Linear Formula | 1·5 H2O2 |
Solubility Information | Solubility in water: 150g/L. |
Formula Weight | 157.01 |
Percent Purity | ≥13% (available oxygen) |
Packaging | Plastic drum |
Chemical Name or Material | Sodium percarbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'