missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-7080605
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
10486-00-7
NaBO3·4H2O
MFCD00149231
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
10486-00-7
NaBO3·4H2O
MFCD00149231

Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium perborate tetrahydrate is considered as a source of active hydrogen used in detergents, cleaning products, laundry detergents and bleaches. It finds application as a tooth bleaching agent utilized for tooth. It has antiseptic properties and acts as disinfectant as well as preservative in eye drops. Further, it is mixed with suitable activator, tetraacetylethylenediamine in order to release oxygen at lower temperatures.

Solubility
Soluble in water.

Notes
Moisture sensitive.Incompatible with metals, metallic salts, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 60°C (decomposition)
Density 1.73
pH ∼10
Odor Odorless
Quantity 100 g
UN Number UN1479
Sensitivity Hygroscopic
Merck Index 14,8652
Solubility Information Soluble in water.
Formula Weight 153.86 (81.84 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium perborate tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'