missing translation for 'onlineSavingsMsg'
Learn More

Sodium pentacyanonitrosylferrate(III) dihydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-4824955
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
13755-38-9
C5H4FeN6Na2O3
297.95
MFCD00149192
JFPDVRGGFBUORC-UHFFFAOYSA-N
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
11953895
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
This item is not returnable. View return policy
13755-38-9
C5H4FeN6Na2O3
297.95
MFCD00149192
JFPDVRGGFBUORC-UHFFFAOYSA-N
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
11953895
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

For Chromatographic Detection of Peptides

For chromatographic detection of peptides.Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.0-102.0%
Solubility Information Soluble in water. Slightly soluble ethanol.
Formula Weight 297.95 (261.92 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.72
Chemical Name or Material Sodium pentacyanonitrosylferrate(III) dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'