missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium oxide, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >400°C (decomposition) |
Physical Form | Beads |
Quantity | 25 g |
UN Number | UN1825 |
Merck Index | 14,8651 |
Solubility Information | Soluble in water. |
Formula Weight | 61.98 |
Sensitivity | Air and moisture sensitive |
Density | 2.27 g/mL |
Chemical Name or Material | Sodium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'