missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Oxalate, Pure, Fisher Chemical™
Click to view available options
Quantity:
500 g
Specifications
Melting Point | 250°C |
Color | White |
pH | 8 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Formula Weight | 134g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium oxalate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'