missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 3801-01

Additional Details : CAS Number : 62-76-0 Weight : 1.00000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 500g
Packaging Poly Bottle

 View more versions of this product

Product Code. 15508174

  • £273.00 / 500g


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

Sodium Oxalate
Titrimetry Standard
NaOCOCOONa
62-76-0
Baker Analyzed™ ACS Reagent
134

Specifications

Sodium Oxalate
62-76-0
Powder
134
1L = 2.34kg
Titrimetry Standard
Baker Analyzed™ ACS Reagent
NaOCOCOONa
Poly Bottle
500g
Videos