missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium orthosilicate, (mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution), Thermo Scientific Chemicals
28.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
Description
It is used as a degreasant, industrial alkaline detergents, water treatment (boiler-feed); adhesives, builder in soaps.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 1 kg |
UN Number | UN1759 |
Solubility Information | Soluble in water. |
Odor | Odorless |
Chemical Name or Material | Sodium orthosilicate, Mixture of NaOH and Na2SiO3 yielding ≈ Na4SiO4 in solution |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'