missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, +97%, For analysis, ACROS Organics™

Brand:  Acros Organics 424350100

Code : 29

Additional Details : CAS Number : 7632-00-0 Weight : 10.05000kg Transport : UN number : 1500 Chem class : 1 Pack group : III

Quantity 10kg
Packaging Plastic Bucket

 View more versions of this product

Product Code. 10258003

  • £336.30 / 10kg


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Sodium nitrite
ACS Reagent Grade
97+%
NaNO2
MFCD00011118
7632-00-0
≥97.0%
NNaO2
69

Specifications

Sodium nitrite
ACS Reagent Grade
≥97.0%
NNaO2
69
01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86
100ppm max.
10ppm max.
10ppm max.
50ppm max.
>320°C
10kg
7632-00-0
Crystals or Crystalline Powder
97+%
NaNO2
MFCD00011118
15, 8779
50ppm max.
0.01% max.
Plastic Bucket
100ppm max.
Light yellow
Videos
SDS