missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, +97%, For analysis, ACROS Organics™

Brand:  Acros Organics 424355000

Code : 29

Additional Details : CAS Number : 7632-00-0 Weight : 0.58000kg Transport : UN number : 1500 Chem class : 5.1 Pack group : III


Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10637961

  • £30.10 / 500g


EU Stock 20
Add to basket


Specifications

Chemical Identifiers

Sodium nitrite
ACS Reagent Grade
97+%
NaNO2
MFCD00011118
7632-00-0
≥97.0%
NNaO2
69

Specifications

Sodium nitrite
ACS Reagent Grade
≥97.0%
NNaO2
69
01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86
100ppm max.
10ppm max.
10ppm max.
50ppm max.
>320°C
500g
7632-00-0
Crystals or Crystalline Powder
97+%
NaNO2
MFCD00011118
15, 8779
50ppm max.
0.01% max.
Plastic bottle
100ppm max.
Light yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium nitrite

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS