missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, TraceSELECT™, ≥99.999% (metals basis), Honeywell™ Fluka™

Catalog Number 15636840
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
This item is not returnable. View return policy
7631-99-4
NNaO3
84.99
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
24268
CHEBI:63005
sodium nitrate
[Na+].[O-][N+]([O-])=O
This item is not returnable. View return policy
7631-99-4
NNaO3
84.99
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
24268
CHEBI:63005
sodium nitrate
[Na+].[O-][N+]([O-])=O
Boiling Point Decomposes on heating
Melting Point 308°C
Color Colorless
pH 5.0 to 7.0 (at 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula NaNO3
UN Number UN1498
Solubility Information 883g/L (at 20°C)
Formula Weight 84.99g/mol
Percent Purity ≥99.999% (metals basis)
Grade TraceSELECT™
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Sodium nitrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'