missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 3770-01

Additional Details : CAS Number : 7631-99-4 Weight : 0.64000kg Transport : UN number : 1498 Chem class : 5.1 Pack group : III

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Product Code. 15588164

  • £187.02 / 500g

Estimated Shipment
21-04-2021

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

7631-99-4
84.994
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[N+](=O)([O-])[O-].[Na+]
NNaO3
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium;nitrate

Specifications

Sodium Nitrate
NNaO3
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
BAKER ANALYZED™ ACS Reagent
1L = 2.26kg
7631-99-4
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
[N+](=O)([O-])[O-].[Na+]
84.994
CHEBI:63005
Crystals
Poly Bottle
500g
Videos
SDS
Documents