missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, 99+%, for biochemistry, ACROS Organics™

Brand:  Acros Organics 446810010

Code : 29

Additional Details : CAS Number : 7631-99-4 Weight : 1.05000kg Transport : UN number : 1498 Chem class : 1 Pack group : III

Quantity 1kg
Packaging Glass bottle

 View more versions of this product

Product Code. 12133761

  • £73.30 / 1kg


EU Stock 4
Add to basket

Specifications

Chemical Identifiers

Sodium nitrate
For biochemistry
NNaO3
84.99
7631-99-4
99+%
NaNO3

Specifications

Sodium nitrate
For biochemistry
99+%
NaNO3
Glass bottle
380.0°C
1kg
7631-99-4
99+%
NNaO3
84.99
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
306.0°C
Videos
SDS