Sodium nitrate, 99+%, for biochemistry, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 446812500

UNSPSC: 12161500

Code: 29

Additional Details:
Additional Details: CAS Number: 7631-99-4 Weight: 0.30000kg Transport: UN number : 1498 Chem class : 1 Pack group : III


Packaging Glass bottle
Quantity 250g

Product Code. 12143761

Quantity Price
1 £ 25.4 / 250g
EU Stock 12
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Sodium nitrate
CAS 7631-99-4
Purity Grade For biochemistry
Assay Percent Range 99+%
Molecular Formula NNaO3
Linear Formula NaNO3
Formula Weight 84.99

Specification

Chemical Name or Material Sodium nitrate
CAS 7631-99-4
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement May intensify fire; oxidizer. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Purity Grade For biochemistry
Assay Percent Range 99+%
Molecular Formula NNaO3
Linear Formula NaNO3
Formula Weight 84.99
Packaging Glass bottle
Quantity 250g
Documentation

Technical documents

Safety and Handling

Warning

GHS P Statement May intensify fire; oxidizer. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.