missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, 99+%, ACS reagent, ACROS Organics™

Brand:  Acros Organics 424340010

Code : 29

Additional Details : CAS Number : 7631-99-4 Weight : 1.05000kg Transport : UN number : 1498 Chem class : 5.1 Pack group : III

Packaging Plastic Bottle
Quantity 1kg

 View more versions of this product

Product Code. 10063973

  • £53.34 / 1kg

Estimated Shipment
09-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7631-99-4
84.99
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
CHEBI:63005
[N+](=O)([O-])[O-].[Na+]
NNaO3
VWDWKYIASSYTQR-UHFFFAOYSA-N
24268
sodium;nitrate

Specifications

Sodium nitrate
99+%
NaNO3
15, 8778
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol
[N+](=O)([O-])[O-].[Na+]
84.99
CHEBI:63005
Crystals
ACS Reagent
10ppm max
5ppm max.
5ppm max.
20ppm max.
Plastic Bottle
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
380.0°C
306.0°C
1kg
7631-99-4
NNaO3
16,86
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium;nitrate
24268
84.99
99+%
50ppm max.
10ppm max.
0.005% max
3ppm max.
10ppm max.
5ppm max.
30ppm max.
White
5.5 to 8.3 (5% soln. at 25°C)
Videos
SDS Documents