missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, +99%, for analysis, ACROS Organics™

Brand:  Acros Organics 205960050

Code : 29

Additional Details : CAS Number : 7631-99-4 Weight : 5.05000kg Transport : UN number : 1498 Chem class : 5.1 Pack group : III

Packaging Plastic bottle
Quantity 5kg

 View more versions of this product

Product Code. 10155643

  • £153.20 / 5kg


Estimated Shipment
10-07-2018
Add to basket

Specifications

Chemical Identifiers

Sodium nitrate
For analysis
99+%
NaNO3
MFCD00011119
7631-99-4
≥99%
NNaO3
84.99

Specifications

Sodium nitrate
For analysis
≥99%
NNaO3
84.99
16,86
50ppm max.
5ppm max.
10ppm max.
5ppm max.
30ppm max.
White
5.5 to 8.3 (5% soln. at 25°C)
7631-99-4
Crystals or Crystalline Powder
99+%
NaNO3
MFCD00011119
15, 8778
10ppm max.
3ppm max.
Plastic bottle
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
380.0°C
306.0°C
5kg
Videos
SDS