missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, +99%, for analysis, ACROS Organics™

Brand:  Acros Organics 205962500

Code : 29

Additional Details : CAS Number : 7631-99-4 Weight : 0.30000kg Transport : UN number : 1498 Chem class : 5.1 Pack group : III


Quantity 250g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10405305

  • £18.00 / 250g


In Stock 4
EU Stock 15
Add to basket


Specifications

Chemical Identifiers

Sodium nitrate
For analysis
99+%
NaNO3
MFCD00011119
7631-99-4
≥99%
NNaO3
84.99

Specifications

Sodium nitrate
For analysis
≥99%
NNaO3
84.99
16,86
50ppm max.
5ppm max.
10ppm max.
5ppm max.
30ppm max.
White
5.5 to 8.3 (5% soln. at 25°C)
7631-99-4
Crystals or Crystalline Powder
99+%
NaNO3
MFCD00011119
15, 8778
10ppm max.
3ppm max.
Plastic bottle
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
380.0°C
306.0°C
250g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium nitrate

Signal Word
  • Warning

Hazard Category
  • Oxidising solid Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P221-Take any precaution to avoid mixing with combustibles .
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

SDS