missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate, 99.999%, (trace metal basis), Acros Organics™

Brand:  Acros Organics 437165000

Code : NEW

Additional Details : CAS Number : 7631-99-4 Weight : 0.50000kg Transport : UN number : 1498 Chem class : 1 Pack group : III

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 13478588

  • £999.10 / 500g


Estimated Shipment
12-02-2018
Add to basket

Specifications

Chemical Identifiers

Sodium nitrate
Trace Metal Basis
NNaO3
84.99
7631-99-4
99.999%
NaNO3

Specifications

Sodium nitrate
Trace Metal Basis
99.999%
NNaO3
84.99
Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
380.0°C
306.0°C
7631-99-4
Crystals
99.999%
NaNO3
Glass bottle
10ppm max.
White
500g
Videos