missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-dodecyl sulfate, 99% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog Number p-7081280
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
151-21-3
C12H25NaO4S
288.38
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
3423265
CHEBI:8984
sodium;dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
This item is not returnable. View return policy
151-21-3
C12H25NaO4S
288.38
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
3423265
CHEBI:8984
sodium;dodecyl sulfate
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

Solubility
Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼206°C
Assay Percent Range 99% (dry wt.)
Quantity 25 g
UN Number UN2926
Beilstein 3599286
Merck Index 148636
Solubility Information Slightly soluble in ethanol.
Formula Weight 288.38
Percent Purity 99%
Assay (dry wt.)
Chemical Name or Material Sodium n-dodecyl sulfate, Water <1.5%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'