missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdenum oxide, anhydrous, Mo 46.2%, Thermo Scientific Chemicals

Catalog Number p-7048960
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7631-95-0
MoNa2O4
205.926
MFCD00003486
TVXXNOYZHKPKGW-UHFFFAOYSA-N
sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
61424
CHEBI:75215
disodium;dioxido(dioxo)molybdenum
[O-][Mo](=O)(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
7631-95-0
MoNa2O4
205.926
MFCD00003486
TVXXNOYZHKPKGW-UHFFFAOYSA-N
sodium molybdate, disodium molybdate, sodium molybdate vi, natriummolybdat, molybdic acid, disodium salt, natriummolybdat german, sodium molybdate van, sodium molybdate na2moo4, unii-948qaq08i1, ccris 5442
61424
CHEBI:75215
disodium;dioxido(dioxo)molybdenum
[O-][Mo](=O)(=O)[O-].[Na+].[Na+]

It is used as an analytical reagent, corrosion inhibitor, and non-oxidizing anodic inhibitor. It reduces the nitrite requirement of fluids inhibited with nitrite-amine and improves the corrosion protection of carboxylate salt fluids. It is utilized in pigments, dyes, anti-freezing agent, cutting oils, metal finishing agent, brightening agent for zinc, additive for fertilizers and micronutrients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 687°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Na2MoO4
Merck Index 14,8645
Solubility Information It is soluble in water.
Formula Weight 205.92
Concentration or Composition (by Analyte or Components) Mo 46.2%
Odor Odorless
Refractive Index 1.714
Sensitivity Hygroscopic
Density 3.72 g/mL
Chemical Name or Material Sodium molybdenum oxide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'