missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methoxide Titrant (0.5M in Methanol), Thermo Scientific Chemicals

Catalog Number 10637391
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
124-41-4, 67-56-1
CH3NaO
54.024
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]
124-41-4, 67-56-1
CH3NaO
54.024
MFCD00012179
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
10942334
sodium;methanolate
C[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 65.0°C
Melting Point -98.0°C
Linear Formula CH3ONa
Concentration 0.45 to 0.60M
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Density 0.8000g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 54.02
Grade Pure
Packaging Glass bottle
Specific Gravity 0.8
Quantity 500 mL
Flash Point 11°C
Physical Form Liquid
Chemical Name or Material Sodium methoxide, 0.5 M in methanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'