missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium methoxide, ca 25% w/w in methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2500 mL
Description
Sodium methoxide 2% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -2°C |
Color | Colorless to Yellow |
Density | 0.945 g/mL |
Boiling Point | 92°C |
Flash Point | 29°C (84°F) |
Linear Formula | NaOCH3 |
Quantity | 2500 mL |
UN Number | UN1289 |
Beilstein | 3592982 |
Sensitivity | Air and moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'