missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metatungstate, 2.82 density solution in water, stabilized, Thermo Scientific Chemicals
365.35 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green to Yellow |
Linear Formula | Na6[H2W12O40] |
Density | 2.8200g/mL |
Solubility Information | Solubility in water: >1000g/L water (20°C). |
Formula Weight | 2986.13 |
Percent Purity | ≥67% (WO3) |
Packaging | Glass bottle |
Specific Gravity | 2.82 |
Quantity | 250 mL |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Sodium metatungstate, 2.82 density solution in water, stabilized
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'