missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaborate tetrahydrate, tech., Thermo Scientific Chemicals

Catalog Number p-7050408
Click to view available options
Quantity:
1 kg
5 kg
10555-76-7
BH8NaO6
137.86
MFCD00149205
JAKYJVJWXKRTSJ-UHFFFAOYSA-N
sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
23694267
sodium;oxido(oxo)borane;tetrahydrate
O.O.O.O.[Na+].[O-]B=O
10555-76-7
BH8NaO6
137.86
MFCD00149205
JAKYJVJWXKRTSJ-UHFFFAOYSA-N
sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
23694267
sodium;oxido(oxo)borane;tetrahydrate
O.O.O.O.[Na+].[O-]B=O

Sodium metaborate tetrahydrate is used In textile industry, it is used as additives, process aid and flame retardant. In chemical industry, it is used to make boric acid. Other uses include thickening agent, preservative and photographic chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼53°C (decomposition)
Physical Form Crystalline
Quantity 1 kg
Solubility Information Soluble in water.
Formula Weight 137.86 (65.80 Anhydrous)
Grade Technical
Chemical Name or Material Sodium metaborate tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'