missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metaborate tetrahydrate, tech., Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
Description
Sodium metaborate tetrahydrate is used In textile industry, it is used as additives, process aid and flame retardant. In chemical industry, it is used to make boric acid. Other uses include thickening agent, preservative and photographic chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼53°C (decomposition) |
Physical Form | Crystalline |
Quantity | 5 kg |
Solubility Information | Soluble in water. |
Formula Weight | 137.86 (65.80 Anhydrous) |
Grade | Technical |
Chemical Name or Material | Sodium metaborate tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'