missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metaborate tetrahydrate, 98.5%, extra pure, Thermo Scientific Chemicals
12.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 57.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 500 g |
Assay Percent Range | 98.5% |
Linear Formula | NaBO2·4H2O |
Merck Index | 10,8477 |
Solubility Information | Solubility in water: 41.9% (20°C). |
IUPAC Name | sodium tetrahydrate oxoborinate |
Formula Weight | 137.86 |
Show More |
Safety and Handling
Product Identifier
- Sodium metaborate tetrahydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'