missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, ACS, 97% min, Thermo Scientific Chemicals

Catalog Number p-4407910
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
7681-57-4
Na2O5S2
190.09
MFCD00167602
LDTLADDKFLAYJA-UHFFFAOYSA-L
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
656671
disodium (sulfinatooxy)sulfinate
[Na+].[Na+].[O-]S(=O)OS([O-])=O
This item is not returnable. View return policy
7681-57-4
Na2O5S2
190.09
MFCD00167602
LDTLADDKFLAYJA-UHFFFAOYSA-L
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
656671
disodium (sulfinatooxy)sulfinate
[Na+].[Na+].[O-]S(=O)OS([O-])=O

Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 97% min.
Solubility Information Soluble in water,ethanol and glycerol.
Formula Weight 190.1
Grade ACS
Odor Pungent
Sensitivity Air and moisture sensitive
Appearance White to pale yellow powder/flake/crystalline/beads
Density 1.48 g/cm3 at 20°C
Chemical Name or Material Sodium metabisulfite, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'