missing translation for 'onlineSavingsMsg'
Learn More

Sodium L-(+)-tartrate dihydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-7023110
Click to view available options
Quantity:
250 g
1000 g
5000 g
6106-24-7
C4H8Na2O8
230.08
MFCD00150035
FGJLAJMGHXGFDE-UDMGOBQRNA-L
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
131855972
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
6106-24-7
C4H8Na2O8
230.08
MFCD00150035
FGJLAJMGHXGFDE-UDMGOBQRNA-L
unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
131855972
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium L-(+)-tartrate dihydrate is used in buffers for molecular biology and cell culture applications. It is also used as an emulsifier and a binding agent in food products such as jellies, margarine, and sausage casingsand also as a food additive.

Solubility
It is soluble in water (290 mg/ml at 20°C). Insoluble in alcohol, and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼150°C (in -2H2O)
Density 1.82
Odor Odorless
Quantity 250 g
Beilstein 6121732
Merck Index 14,8684
Solubility Information It is soluble in water (290mg/ml at 20°C). Insoluble in alcohol,and organic solvents.
Formula Weight 230.08 (194.06 Anhydrous)
Percent Purity 99%
Chemical Name or Material Sodium L-(+)-tartrate dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'