missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049257
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7681-82-5
INa
149.894
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]
This item is not returnable. View return policy
7681-82-5
INa
149.894
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]

Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of alkyl chlorides into alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1300°C to 1304°C
Melting Point 651°C
Color White
Physical Form Beads
Quantity 5 g
Merck Index 14,8631
Solubility Information Very soluble in water and liquid ammonia. Freely soluble in acetone,formic acid,formamide,methanol,and acetonitrile. Soluble in N,N-dimethylformamide.
Formula Weight 149.89
Percent Purity 99.999%
Odor Odorless
Packaging Ampule
Refractive Index 1.7745
Sensitivity Hygroscopic; Light sensitive
Density 3.67 g/mL
Assay (metals basis)
Chemical Name or Material Sodium iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'