missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodate, 99% min, Thermo Scientific Chemicals

Catalog Number p-7050974
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7681-55-2
INaO3
197.891
MFCD00003533
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
23675764
CHEBI:81708
sodium;iodate
[O-]I(=O)=O.[Na+]
This item is not returnable. View return policy
7681-55-2
INaO3
197.891
MFCD00003533
WTCBONOLBHEDIL-UHFFFAOYSA-M
sodium iodate, natriumjodat german, iodic acid, sodium salt, sodium iodate naio3, iodic acid hio3 , sodium salt, sodium iodate naio3 pentahydrate, unii-u558pcs5z9, hsdb 749, iodic acid, sodium salt, pentahydrate, natriumjodat
23675764
CHEBI:81708
sodium;iodate
[O-]I(=O)=O.[Na+]

Sodium iodate is used in animal feed, food industry. It is utilized in the manufacture of disinfectants, antiseptics and deodorants. It acts as a significant part in the oxidation of aromatic, aliphatic, and alicyclic thiols. Potassium tetrahydroborate and sodium iodate in acidic aqueous solution react with cadmium to generate chemical vapor, which is useful for atomic fluorescence spectrometry application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color White
Physical Form Crystalline Powder
Quantity 100 g
UN Number UN1479
Merck Index 14,8630
Solubility Information Soluble in water and acetone.
Formula Weight 197.89
Percent Purity ≤99%
Sensitivity Light and moisture sensitive
Density 4.28 g/mL
Chemical Name or Material Sodium iodate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'