Learn More
Sodium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Specification Of pH.Eur., Bp, Fisher Chemical™
HNaO, CAS Number-1310-73-2, sodium hydroxide solution, ascarite, aetznatron, soda lye, sodium hydrate, caustic soda, sodium hydroxide na oh, sodium hydroxide, white caustic, soda, caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Brand: Fisher Chemical J/7620/15
Code : 23
Additional Details : CAS Number : 1310-73-2 Weight : 1.04000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Description
HNaO, CAS Number-1310-73-2, sodium hydroxide solution, ascarite, aetznatron, soda lye, sodium hydrate, caustic soda, sodium hydroxide na oh, sodium hydroxide, white caustic, soda, caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BPSpecifications
Colorless | |
1310-73-2 | |
3548 | |
1 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
23 &20 |
100°C | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
HDPE plastic bottle | |
Liquid |