missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 Ampule
6 Ampules
Description
HNaO, CAS Number-1310-73-2, sodium hydroxide, sodium hydroxide na oh, soda lye, caustic soda, ascarite, white caustic, sodium hydrate, sodium hydroxide solution, soda, caustic, aetznatron, 6 ampules, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997
Specifications
Color | Colorless |
Boiling Point | 100°C |
Quantity | 6 Ampules |
Packaging | Ampoule |
Vapor Pressure | 23 &20 |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'