missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, purum p.a, >97%, Honeywell™
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
pH | Alkaline |
Color | Colorless |
Boiling Point | 1390°C |
Melting Point | 319°C |
Linear Formula | NaOH |
UN Number | UN1823 |
Density | 2.13g/cm3 |
Quantity | 25 kg |
Formula Weight | 40g/mol |
Packaging | Poly bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'