missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide NF/FCC Pellet, Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly
Specifications
Quantity | 500 g |
Formula Weight | 40 |
Grade | FCC/NF |
Packaging | POLYSTORMOR |
Physical Form | Pellet |
Chemical Name or Material | Sodium Hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'