missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Free-flowing, ACS reagent, anhydrous, pellets, ≥97%, Honeywell Fluka™

Catalog Number p-7111992
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

Free-flowing, ACS reagent, anhydrous, pellets,≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1,390°C
Melting Point 318°C (literature)
CAS Max % 100.0000%
Linear Formula NaOH
UN Number UN1823
Density 2.13 g/cm3
Quantity 500 g
Formula Weight 40.00
Percent Purity >97%
Grade ACS Reagent
Packaging Poly Bottle
Physical Form Pellet
Chemical Name or Material Sodium hydroxide, Free-flowing, ACS reagent, anhydrous, pellets, ≥97%
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'