missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
41.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
41.25 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
41.25 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Description
HNaO, CAS Number-1310-73-2, white caustic, aetznatron, sodium hydroxide na oh, caustic soda, ascarite, soda lye, soda, caustic, sodium hydroxide solution, sodium hydrate, sodium hydroxide, 5kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548
Specifications
pH | 14 |
Color | White |
Boiling Point | 1390°C |
Melting Point | 318°C |
Quantity | 500 g |
Formula Weight | 40g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1mbar 700 |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'