missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | NaOH |
UN Number | 1823 |
Solubility Information | Soluble in water, 1260 g/L at 20°C |
Quantity | 25 g |
Formula Weight | 40g/mol |
Percent Purity | ≥97% |
Grade | ACS Reagent |
Vapor Density | >1 (vs air) |
Vapor Pressure | 18mmHg (20°C), 3mmHg (37°C) |
Chemical Name or Material | Sodium Hydroxide |
Safety and Handling
Product Identifier
- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'