missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.5N, Thermo Scientific Chemicals

Catalog Number 11319329
Click to view available options
Quantity:
1 U
6 U
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
This item is not returnable. View return policy
1310-73-2
HNaO
39.997
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN1824
Formula Weight 40
Odor Odorless
Quantity 1 U
Physical Form Liquid
Chemical Name or Material Sodium hydroxide Acculute Standard Volumetric Solution, Final Concentration 0.5N

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'